• USA Home
  • V900158 - Pyrophosphoric acid

EMAIL THIS PAGE TO A FRIEND
V900158 Sigma-Aldrich

Pyrophosphoric acid

Vetec reagent grade, 94%

Synonym: Diphosphoric acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Inorganic Acids, Synthetic Reagents More...
grade   Vetec reagent grade
InChI Key   XPPKVPWEQAFLFU-UHFFFAOYSA-N
assay   94%

Description

Packaging

100 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?