EMAIL THIS PAGE TO A FRIEND
V900263 Sigma-Aldrich

Benzaldehyde

Vetec, reagent grade, 98%

Synonym: Bitter almond

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7, Carbonyl Compounds, Chemical Synthesis,
grade   reagent grade
vapor density   3.7 (vs air)
vapor pressure   4 mmHg ( 45 °C)
product line   Vetec
assay   98%
form   liquid
autoignition temp.   374 °F
expl. lim.   1.4 %, 20 °F
refractive index   n20/D 1.545 (lit.)
pH   5.9 (20 °C)
bp   178-179 °C (lit.)
mp   −26 °C (lit.)
density   1.044 g/cm3 at 20 °C (lit.)
SMILES string   O=Cc1ccccc1
InChI   1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H
InChI key   HUMNYLRZRPPJDN-UHFFFAOYSA-N

Description

Packaging

500 mL in glass bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1990 9 / PGIII
WGK Germany 
WGK 1
Flash Point(F) 
145.4 °F - closed cup
Flash Point(C) 
63 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?