EMAIL THIS PAGE TO A FRIEND
V900434 Sigma-Aldrich

D-Threonine

Vetec, reagent grade, ≥98%

Synonym: (2R,3S)-2-Amino-3-hydroxybutyric acid, D-α-Amino-β-hydroxybutyric acid

  • CAS Number 632-20-2

  • Empirical Formula (Hill Notation) C4H9NO3

  • Molecular Weight 119.12

  •  Beilstein/REAXYS Number 1721643

  •  EC Number 211-171-8

  •  MDL number MFCD00064269

  •  PubChem Substance ID 329829840

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
grade   reagent grade
product line   Vetec
assay   ≥98%
form   powder
color   white
mp   274 °C
SMILES string   C[C@H](O)[C@@H](N)C(O)=O
InChI   1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m0/s1
InChI key   AYFVYJQAPQTCCC-STHAYSLISA-N

Description

Packaging

5, 25 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Threonine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Threonine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Keywords: Chromatography, High performance liquid chromatography

Related Content

Amino Acids Reference Charts

Amino acids are the compounds or building blocks that make up peptides and proteins. Each amino acid is structured from an amino group and a carboxyl group bound to a tetrahedral carbon. This carbon ...
Keywords: Building blocks, Gene expression, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?