EMAIL THIS PAGE TO A FRIEND
V900440 Sigma-Aldrich

BES sodium salt

Vetec, reagent grade, 99%

Synonym: N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid sodium salt

Purchase

Properties

grade   reagent grade
product line   Vetec
assay   99%
form   crystalline powder
useful pH range   6.4 - 7.8
pKa (25 °C)   7.1 (Free acid)
solubility   water: 333.3 mg/mL, clear, colorless
SMILES string   [Na+].OCCN(CCO)CCS([O-])(=O)=O
InChI   1S/C6H15NO5S.Na/c8-4-1-7(2-5-9)3-6-13(10,11)12;/h8-9H,1-6H2,(H,10,11,12);/q;+1/p-1
InChI key   CFQLQLSIZOWFNV-UHFFFAOYSA-M

Description

Packaging

25, 100 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Buffers - From Bnech to Bulk
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?