EMAIL THIS PAGE TO A FRIEND
V900441 Sigma-Aldrich

D-Phenylalanine

Vetec, reagent grade, ≥98%

Synonym: (R)-2-Amino-3-phenylpropionic acid

  • CAS Number 673-06-3

  • Empirical Formula (Hill Notation) C9H11NO2

  • Molecular Weight 165.19

  •  Beilstein/REAXYS Number 2804068

  •  EC Number 211-603-5

  •  MDL number MFCD00004270

  •  PubChem Substance ID 329829847

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine More...
grade   reagent grade
product line   Vetec
assay   ≥98%
form   powder
color   white to off-white
mp   273-276 °C
SMILES string   N[C@H](Cc1ccccc1)C(O)=O
InChI   1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m1/s1
InChI key   COLNVLDHVKWLRT-MRVPVSSYSA-N

Description

Packaging

5, 25 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?