EMAIL THIS PAGE TO A FRIEND
V900466 Sigma-Aldrich

L-Threonine

Vetec, reagent grade, ≥98%

Synonym: (2S,3R)-2-Amino-3-hydroxybutyric acid

  • CAS Number 72-19-5

  • Linear Formula CH3CH(OH)CH(NH2)CO2H

  • Molecular Weight 119.12

  •  Beilstein/REAXYS Number 1721646

  •  EC Number 200-774-1

  •  MDL number MFCD00064270

  •  PubChem Substance ID 329829867

Purchase

Properties

grade   reagent grade
product line   Vetec
assay   ≥98%
form   powder
color   white to off-white
mp   256 °C (dec.) (lit.)
SMILES string   C[C@@H](O)[C@H](N)C(O)=O
InChI   1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1
InChI key   AYFVYJQAPQTCCC-GBXIJSLDSA-N

Description

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?