• USA Home
  • V900469 - D-Lysine monohydrochloride

EMAIL THIS PAGE TO A FRIEND
V900469 Sigma-Aldrich

D-Lysine monohydrochloride

Vetec, reagent grade, ≥98%

Synonym: (R)-2,6-Diaminohexanoic acid monohydrochloride

  • CAS Number 7274-88-6

  • Linear Formula H2N(CH2)4CH(NH2)CO2H·HCl

  • Molecular Weight 182.65

  •  Beilstein/REAXYS Number 4356907

  •  MDL number MFCD00012920

  •  PubChem Substance ID 329829870

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
grade   reagent grade
product line   Vetec
assay   ≥98%
form   powder
color   white to off-white
mp   266 °C (dec.) (lit.)
SMILES string   Cl[H].NCCCC[C@@H](N)C(O)=O
InChI   1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m1./s1
InChI key   BVHLGVCQOALMSV-NUBCRITNSA-N

Description

Packaging

25 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Amino Acids Reference Charts

Amino acids are the compounds or building blocks that make up peptides and proteins. Each amino acid is structured from an amino group and a carboxyl group bound to a tetrahedral carbon. This carbon ...
Keywords: Building blocks, Gene expression, Mass spectrometry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?