EMAIL THIS PAGE TO A FRIEND
V900664 Sigma-Aldrich

1,3-Propanediol

Vetec, reagent grade, 98%

Synonym: 1,3-Dihydroxypropane, Bio-PDO, PG, Trimethylene glycol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
grade   reagent grade
vapor pressure   0.8 mmHg ( 20 °C)
  9.8 mmHg ( 100 °C)
product line   Vetec
assay   98%
refractive index   n20/D 1.440 (lit.)
bp   214 °C/760 mmHg (lit.)
mp   −27 °C (lit.)
density   1.053 g/mL at 25 °C (lit.)
SMILES string   OCCCO
InChI   1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2
InChI key   YPFDHNVEDLHUCE-UHFFFAOYSA-N

Description

Packaging

500 mL in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
>210.2 °F
Flash Point(C) 
>99 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?