EMAIL THIS PAGE TO A FRIEND
V900684 Sigma-Aldrich

DL-Tryptophan

Vetec, reagent grade, 98%

Synonym: (±)-α-Amino-3-indolepropionic acid, (±)-2-Amino-3-(3-indolyl)propionic acid, DL-3β-Indolylalanine

  • CAS Number 54-12-6

  • Empirical Formula (Hill Notation) C11H12N2O2

  • Molecular Weight 204.23

  •  Beilstein/REAXYS Number 86199

  •  EC Number 200-194-9

  •  MDL number MFCD00064339

  •  PubChem Substance ID 329830056

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Tryptophan More...
grade   reagent grade
product line   Vetec
assay   98%
form   powder
color   white to off-white
mp   289-290 °C (dec.) (lit.)
SMILES string   NC(Cc1c[nH]c2ccccc12)C(O)=O
InChI   1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)
InChI key   QIVBCDIJIAJPQS-UHFFFAOYSA-N

Description

Packaging

5, 25 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?