EMAIL THIS PAGE TO A FRIEND
V900711 Sigma-Aldrich

Dulcitol

Vetec, reagent grade, 98%

Synonym: Dulcite, Galactitol

  • CAS Number 608-66-2

  • Empirical Formula (Hill Notation) C6H14O6

  • Molecular Weight 182.17

  •  Beilstein/REAXYS Number 1721903

  •  EC Number 210-165-2

  •  MDL number MFCD00064288

  •  PubChem Substance ID 329830083

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F
grade   reagent grade
product line   Vetec
assay   98%
SMILES string   OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)CO
InChI   1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5+,6-
InChI key   FBPFZTCFMRRESA-GUCUJZIJSA-N

Description

Packaging

25, 100 g in poly bottle

Biochem/physiol Actions

A sugar alcohol and slightly sweet tastant for human taste cells.

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?