EMAIL THIS PAGE TO A FRIEND
V900720 Sigma-Aldrich

DL-Aspartic acid

Vetec, reagent grade, 98%

Synonym: (±)-2-Aminosuccinic acid

  • CAS Number 617-45-8

  • Linear Formula HOOCCH2CH(NH2)COOH

  • Molecular Weight 133.10

  •  Beilstein/REAXYS Number 774618

  •  EC Number 210-513-3

  •  MDL number MFCD00063083

  •  PubChem Substance ID 329830092

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Aspartic Acid, Biochemicals and Reagents, Chemical Biology,
grade   reagent grade
product line   Vetec
assay   98%
form   powder
color   white
mp   >300 °C (lit.)
SMILES string   NC(CC(O)=O)C(O)=O
InChI   1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)
InChI key   CKLJMWTZIZZHCS-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Aspartic Acid Enantiomers on Astec® CLC-L, Trace Level Detection

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Aspartic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
Keywords: Chromatography, High performance liquid chromatography

Related Content

Amino Acids Reference Charts

Amino acids are the compounds or building blocks that make up peptides and proteins. Each amino acid is structured from an amino group and a carboxyl group bound to a tetrahedral carbon. This carbon ...
Keywords: Building blocks, Gene expression, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?