• USA Home
  • V900748 - 4-Chlorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
V900748 Sigma-Aldrich

4-Chlorobenzoic acid

Vetec, reagent grade, 98%

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
grade   reagent grade
product line   Vetec
assay   98%
mp   238-241 °C (lit.)
SMILES string   OC(=O)c1ccc(Cl)cc1
InChI   1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
InChI key   XRHGYUZYPHTUJZ-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
460.4 °F - closed cup
Flash Point(C) 
238 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?