• USA Home
  • V900880 - Sodium thiosulfate pentahydrate

EMAIL THIS PAGE TO A FRIEND
V900880 Sigma-Aldrich

Sodium thiosulfate pentahydrate

Vetec reagent grade

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
grade   Vetec reagent grade
InChI Key   PODWXQQNRWNDGD-UHFFFAOYSA-L

Description

Packaging

500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?