• USA Home
  • 07856 - 4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl)benzylamine

EMAIL THIS PAGE TO A FRIEND
07856 Sigma-Aldrich

4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl)benzylamine

≥95%

Synonym: 4-(1H,1H,2H,2H-Perfluorodecyl)benzylamine

  • CAS Number 609816-23-1

  • Empirical Formula (Hill Notation) C17H12F17N

  • Molecular Weight 553.26

  •  MDL number MFCD03788348

  •  PubChem Substance ID 57646777

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Solution-Phase Scavengers, Specialty Synthesis
Quality Level   100
assay   ≥95%
mp   57-63 °C
SMILES string   NCc1ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc1
InChI   1S/C17H12F17N/c18-10(19,6-5-8-1-3-9(7-35)4-2-8)11(20,21)12(22,23)13(24,25)14(26,27)15(28,29)16(30,31)17(32,33)34/h1-4H,5-7,35H2
InChI key   MNOIHTZEQBURKA-UHFFFAOYSA-N

Description

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?