• USA Home
  • 08686 - 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanethiol

EMAIL THIS PAGE TO A FRIEND
08686 Sigma-Aldrich

3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanethiol

≥96.0%

Synonym: 1H,1H,2H,2H-Perfluoro-1-decanethiol

  • CAS Number 34143-74-3

  • Linear Formula CF3(CF2)7CH2CH2SH

  • Molecular Weight 480.18

  •  MDL number MFCD00792429

  •  PubChem Substance ID 57646779

  •  NACRES NA.23

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Solution-Phase Scavengers, Specialty Synthesis
Quality Level   100
assay   ≥96.0%
refractive index   n20/D 1.333
bp   82 °C/12 mmHg
mp   15 °C
density   1.678 g/mL at 25 °C
SMILES string   FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCS
InChI   1S/C10H5F17S/c11-3(12,1-2-28)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h28H,1-2H2
InChI key   URJIJZCEKHSLHA-UHFFFAOYSA-N

Description

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?