EMAIL THIS PAGE TO A FRIEND
100404 Sigma-Aldrich

DL-Ethionine

99%

Synonym: DL-2-Amino-4-(ethylthio)butyric acid

  • CAS Number 67-21-0

  • Linear Formula C2H5SCH2CH2CH(NH2)COOH

  • Molecular Weight 163.24

  •  Beilstein/REAXYS Number 1722529

  •  MDL number MFCD00063102

Purchase

Properties

assay   99%
SMILES string   CCSCCC(N)C(O)=O
InChI   1S/C6H13NO2S/c1-2-10-4-3-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)
InChI key   GGLZPLKKBSSKCX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?