• USA Home
  • 102318 - 4,4′-Dimethyloctafluorobiphenyl

EMAIL THIS PAGE TO A FRIEND
102318 Aldrich

4,4′-Dimethyloctafluorobiphenyl

97%

Synonym: Octafluoro-p,p′-bitolyl

  • CAS Number 26475-18-3

  • Empirical Formula (Hill Notation) C14H6F8

  • Molecular Weight 326.18

  •  MDL number MFCD00000314

Purchase

Properties

assay   97%
SMILES string   Cc1c(F)c(F)c(c(F)c1F)-c2c(F)c(F)c(C)c(F)c2F
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?