• USA Home
  • 102539 - N-Benzyl-N-ethyl-m-toluidine

EMAIL THIS PAGE TO A FRIEND
102539 Sigma-Aldrich

N-Benzyl-N-ethyl-m-toluidine

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.5870 (lit.)
bp   161-162 °C (lit.)
density   0.770 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2753 6.1 / PGIII
WGK Germany 
2
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?