• USA Home
  • 103225 - Magnesium trifluoroacetylacetonate hydrate

EMAIL THIS PAGE TO A FRIEND
103225 Aldrich

Magnesium trifluoroacetylacetonate hydrate

  • CAS Number 652154-06-8

  • Linear Formula Mg(C5H4F3O2)2 · xH2O

  • Molecular Weight 330.46 (anhydrous basis)

  •  MDL number MFCD00150844

Purchase
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?