• USA Home
  • 104310 - 3,4,5,6-Tetrabromophthalimide

EMAIL THIS PAGE TO A FRIEND
104310

3,4,5,6-Tetrabromophthalimide

97%

Synonym: 4,5,6,7-Tetrabromoisoindole-1,3-dione

Purchase

Properties

assay   97%
SMILES string   Brc1c(Br)c(Br)c2C(=O)NC(=O)c2c1Br
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?