EMAIL THIS PAGE TO A FRIEND
10893 Sigma-Aldrich

Allyl acetoacetate

produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5% (GC)

Synonym: AC-allyl

  • CAS Number 1118-84-9

  • Linear Formula CH3COCH2COOCH2CH=CH2

  • Molecular Weight 142.15

  •  Beilstein/REAXYS Number 1758569

  •  EC Number 214-269-9

  •  MDL number MFCD00009811

  •  PubChem Substance ID 24846840

Purchase

Properties

grade   produced by Wacker Chemie AG, Burghausen, Germany
assay   ≥98.5% (GC)
refractive index   n20/D 1.439 (lit.)
bp   194-195 °C/737 mmHg (lit.)
density   1.037 g/mL at 25 °C (lit.)
SMILES string   CC(=O)CC(=O)OCC=C
InChI   1S/C7H10O3/c1-3-4-10-7(9)5-6(2)8/h3H,1,4-5H2,2H3
InChI key   AXLMPTNTPOWPLT-UHFFFAOYSA-N

Description

Other Notes

prices for bulk quantities on request

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
1
Flash Point(F) 
152.6 °F
Flash Point(C) 
67 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?