• USA Home
  • 10980 - Arecoline hydrobromide

EMAIL THIS PAGE TO A FRIEND
10980 Aldrich

Arecoline hydrobromide

≥99.0% (NT)

Synonym: 1-Methyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid methyl ester hydrobromide, Arecaidine methyl ester hydrobromide, Methyl 1,2,5,6-tetrahydro-1-methyl-3-pyridinecarboxylate hydrobromide, Methyl 1-methyl-1,2,5,6-tetrahydronicotinate hydrobromide, Taeniolin

  • CAS Number 300-08-3

  • Empirical Formula (Hill Notation) C8H13NO2 · HBr

  • Molecular Weight 236.11

  •  Beilstein Registry Number 3914826

  •  EC Number 206-087-3

  •  MDL number MFCD00039041

  •  PubChem Substance ID 24846938

Purchase

Properties

InChI Key   AXOJRQLKMVSHHZ-UHFFFAOYSA-N
assay   ≥99.0% (NT)
mp   170-175 °C
  172-176 °C (lit.)
solubility   H2O: 0.1 g/mL, clear, colorless

Description

Other Notes

Alkaloid from seeds of betel nut palm. Effect on human learning

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QT2275000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?