EMAIL THIS PAGE TO A FRIEND
110361 Sigma-Aldrich

3-Buten-1-ol

≥98%

Synonym: Allylcarbinol

  • CAS Number 627-27-0

  • Linear Formula CH2=CHCH2CH2OH

  • Molecular Weight 72.11

  •  Beilstein/REAXYS Number 1633504

  •  EC Number 210-991-3

  •  MDL number MFCD00002959

Purchase

Properties

InChI Key   ZSPTYLOMNJNZNG-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.421 (lit.)
bp   112-114 °C(lit.)
density   0.838 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
91.4 °F
Flash Point(C) 
33 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

496839 3-Buten-1-ol, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?