EMAIL THIS PAGE TO A FRIEND
112593 Aldrich

D-(+)-Galactose

97%

Synonym: Galactose

  • CAS Number 59-23-4

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 1724619

  •  EC Number 200-416-4

  •  MDL number MFCD00151230

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   GZCGUPFRVQAUEE-KCDKBNATSA-N
assay   97%
optical activity   [α]20/D +80.2°, c = 2 in H2O
mp   168-170 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LW5490000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G0625 D-(+)-Galactose, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?