• USA Home
  • 114731 - Ammonium disodium amminepentacyanoferrate(II) hydrate

EMAIL THIS PAGE TO A FRIEND
114731 Sigma-Aldrich

Ammonium disodium amminepentacyanoferrate(II) hydrate

90%

Purchase

Properties

assay   90%
SMILES string   N.[NH4+].O.[Na+].[Na+].N#C[Fe-3](C#N)(C#N)(C#N)C#N

Description

Other Notes

Contains sodium amminepentacyanoferrate(II) hydrate

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?