• USA Home
  • 115185 - (−)-6-Azauridine 2′,3′,5′-triacetate

EMAIL THIS PAGE TO A FRIEND
115185 Sigma-Aldrich

(−)-6-Azauridine 2′,3′,5′-triacetate

Synonym: 2′,3′,5′-Tri-O-acetyl-6-azauridine, 2-β-D-Ribofuranosyl-1,2,4-triazine-3,5(2H,4H)-dione 2′,3′,5′-triacetate, 6-AzUrd-TA, 6-Azauridine 2′,3′,5′-triacetate, Azaribine

  • CAS Number 2169-64-4

  • Empirical Formula (Hill Notation) C14H17N3O9

  • Molecular Weight 371.30

  •  Beilstein/REAXYS Number 631776

  •  EC Number 218-515-6

  •  MDL number MFCD00006471

  •  PubChem Substance ID 24847203

Purchase

Properties

optical activity   [α]22/D −40.9°, c = 1 in pyridine
mp   99-101 °C (lit.)
SMILES string   CC(=O)OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H]1OC(C)=O)N2N=CC(=O)NC2=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
XY8577000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?