• USA Home
  • 115223 - 3-Chlorobenzyl chloride

EMAIL THIS PAGE TO A FRIEND
115223 Sigma-Aldrich

3-Chlorobenzyl chloride

technical grade

Synonym: 3,α-Dichlorotoluene

  • CAS Number 620-20-2

  • Linear Formula ClC6H4CH2Cl

  • Molecular Weight 161.03

  •  Beilstein/REAXYS Number 742266

  •  EC Number 210-629-4

  •  MDL number MFCD00000905

Purchase

Properties

grade   technical grade
refractive index   n20/D 1.555 (lit.)
bp   215-216 °C (lit.)
density   1.27 g/mL at 25 °C (lit.)
SMILES string   ClCc1cccc(Cl)c1
InChI   1S/C7H6Cl2/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2
InChI key   DDGRAFHHXYIQQR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2235 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?