• USA Home
  • 115657 - S-(4-Chlorobenzyl)thiuronium chloride

EMAIL THIS PAGE TO A FRIEND
115657

S-(4-Chlorobenzyl)thiuronium chloride

≥99%

Synonym: 2-(4-Chlorobenzyl)-2-thiopseudourea hydrochloride

  • CAS Number 544-47-8

  • Empirical Formula (Hill Notation) C8H10Cl2N2S

  • Molecular Weight 237.15

  •  MDL number MFCD00012584

Purchase

Properties

assay   ≥99%
SMILES string   Cl.NC(=N)SCc1ccc(Cl)cc1
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?