• USA Home
  • 116211 - 3-Ethoxy-2-nitropyridine

EMAIL THIS PAGE TO A FRIEND
116211 Sigma-Aldrich

3-Ethoxy-2-nitropyridine

Purchase

Properties

bp   175 °C/50 mmHg(lit.)
mp   30-31 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
UT3001500
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?