• USA Home
  • 116343 - α-Acetyl-α-methyl-γ-butyrolactone

EMAIL THIS PAGE TO A FRIEND
116343 Sigma-Aldrich

α-Acetyl-α-methyl-γ-butyrolactone

95%

Purchase

Properties

assay   95%
refractive index   n20/D 1.456 (lit.)
density   1.155 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?