• USA Home
  • 116823 - Acetoxytriethyllead(IV)

EMAIL THIS PAGE TO A FRIEND
116823 Sigma-Aldrich

Acetoxytriethyllead(IV)

97%

Purchase

Properties

assay   97%
mp   155 °C (dec.) (lit.)
SMILES string   CC[Pb](CC)(CC)OC(C)=O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
3
RTECS 
TP3922000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?