EMAIL THIS PAGE TO A FRIEND
11925 Sigma-Aldrich

Samarium

pieces, purified by sublimation, puriss., 99.9%

Purchase

Properties

grade   puriss.
assay   99.9%
form   pieces
purified by   sublimation
extent of labeling   103.6 Bq/g specific activity
resistivity   91.4 μΩ-cm, 0°C
impurities   ≤0.05% rare earths
  ≤0.1% other elements
bp   1794 °C (lit.)
mp   1074 °C (lit.)
density   7.47 g/mL at 25 °C (lit.)
SMILES string   [Sm]
InChI   1S/Sm
InChI key   KZUNJOHGWZRPMI-UHFFFAOYSA-N

Description

Other Notes

Pertains only in Germany: Die Abgabe radioaktiver Verbindungen ist in der Schweiz bewilligungspflichtig. Wir bitten unsere Schweizer Kunden bei Bestellung um Angabe ihrer Bewilligungsnummer, ausgestellt vom Bundesamt für Gesundheitswesen.

Preparation of samarium diiodide with ethylene iodide used in various reations

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2910 7
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?