• USA Home
  • 119938 - Benzyl chloroformate

EMAIL THIS PAGE TO A FRIEND
119938 Aldrich

Benzyl chloroformate

≥98%

Synonym: Carbobenzoxy chloride, Z-Chloride

  • CAS Number 501-53-1

  • Linear Formula ClCO2CH2C6H5

  • Molecular Weight 170.59

  •  Beilstein Registry Number 509751

  •  MDL number MFCD00000640

  •  PubChem Substance ID 24847303

Purchase

Properties

Related Categories Amine Protection, Chemical Biology, Chemical Synthesis, Other Protecting and Derivatizing Reagents, Peptide Chemistry,
vapor density   1 (vs air)
vapor pressure   1.39 psi ( 20 °C)
InChI Key   HSDAJNMJOMSNEV-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.519(lit.)
bp   103 °C/20 mmHg(lit.)
density   1.195 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Cbz-protected anilines were prepared directly from aromatic carboxylic acids, sodium azide and Cbz-Cl. Also used in a recently reported 3-step synthesis of 1,2,4-oxadiazoles.

Protecting reagent in peptide synthesis.

Useful for the introduction of the benzyloxycarbonyl(Cbz,Z) protecting group

Packaging

2.5 kg in Sure/Seal™

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1739 8 / PGI
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C

Same Products - New Packaging

Screeing simplified
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?