EMAIL THIS PAGE TO A FRIEND
12001 Sigma-Aldrich

Calcium

granular, ≥98% (complexometric)

Synonym: Calcium atom, Calcium element

  • CAS Number 7440-70-2

  • Empirical Formula (Hill Notation) Ca

  • Molecular Weight 40.08

  •  Beilstein/REAXYS Number 4241647

  •  EC Number 231-179-5

  •  MDL number MFCD00085314

Purchase

Properties

assay   ≥98% (complexometric)
quality   granular
resistivity   3.5 μΩ-cm, 20°C
impurities   ≤0.05% insoluble in HCl
particle size   0.2-3 mm
bp   1484 °C (lit.)
mp   850 °C (lit.)
density   1.54 g/mL at 25 °C (lit.)
SMILES string   [Ca]
InChI   1S/Ca
InChI key   OYPRJOBELJOOCE-UHFFFAOYSA-N

Description

Application

Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7

Packaging

bottled under inert gas

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1401 4.3 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?