EMAIL THIS PAGE TO A FRIEND
121398 Sigma-Aldrich

Giemsa stain

certified by the Biological Stain Commission

Synonym: Azure mixture sicc. Giemsa stain

  • CAS Number 51811-82-6

  • Empirical Formula (Hill Notation) C14H14ClN3S

  • Molecular Weight 291.80

  •  MDL number MFCD00081642

  •  PubChem Substance ID 24847458

Purchase

Properties

grade   certified by the Biological Stain Commission
mp   300 °C (lit.)
λmax   521 nm
  648 nm (2nd)
SMILES string   [Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1
InChI   1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H
InChI key   NALREUIWICQLPS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G5637 Giemsa stain, certified by the Biological Stain Commission

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?