• USA Home
  • 121843 - N-Hydroxynaphthalimide sodium salt

EMAIL THIS PAGE TO A FRIEND
121843 Sigma-Aldrich

N-Hydroxynaphthalimide sodium salt

97%

Synonym: N,N-(1,8-Naphthalyl)hydroxylamine sodium salt

  • CAS Number 6207-89-2

  • Empirical Formula (Hill Notation) C12H6NNaO3

  • Molecular Weight 235.17

  •  Beilstein/REAXYS Number 4282212

  •  MDL number MFCD00006923

  •  PubChem Substance ID 24847468

Purchase

Properties

assay   97%
SMILES string   [Na+].[O-]N1C(=O)c2cccc3cccc(C1=O)c23
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?