• USA Home
  • 121894 - Neocuproine hydrochloride hydrate

EMAIL THIS PAGE TO A FRIEND
121894 Sigma-Aldrich

Neocuproine hydrochloride hydrate

98%

Synonym: 2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate

  • CAS Number 332360-00-6

  • Empirical Formula (Hill Notation) C14H12N2 · HCl · xH2O

  • Molecular Weight 244.72 (anhydrous basis)

  •  EC Number 230-732-8

  •  MDL number MFCD00150062

  •  PubChem Substance ID 24847469

Purchase

Properties

assay   98%
mp   250 °C (dec.) (lit.)
SMILES string   O.Cl.Cc1ccc2ccc3ccc(C)nc3c2n1
InChI   1S/C14H12N2.ClH.H2O/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9;;/h3-8H,1-2H3;1H;1H2
InChI key   APDYLFLZNGECIM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N1626 Neocuproine hydrochloride hydrate, BioReagent, crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?