EMAIL THIS PAGE TO A FRIEND
122637 Sigma-Aldrich

Acetamide

≥98%

Synonym: Amide C2

Purchase

Properties

vapor pressure   1 mmHg ( 65 °C)
InChI Key   DLFVBJFMPXGRIB-UHFFFAOYSA-N
assay   ≥98%
bp   221 °C(lit.)
mp   78-80 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
RTECS 
AB4025000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A0500 Acetamide, ~99% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?