EMAIL THIS PAGE TO A FRIEND
125776 Sigma-Aldrich

Decanal

95%

Synonym: Caprinaldehyde, Decyl aldehyde

  • CAS Number 112-31-2

  • Linear Formula CH3(CH2)8CHO

  • Molecular Weight 156.27

  •  Beilstein/REAXYS Number 1362530

  •  EC Number 203-957-4

  •  MDL number MFCD00007031

Purchase

Properties

vapor density   >1 (vs air)
vapor pressure   ~0.15 mmHg ( 20 °C)
assay   95%
refractive index   n20/D 1.428 (lit.)
bp   207-209 °C (lit.)
density   0.83 g/mL at 20 °C
  0.83 g/mL at 25 °C (lit.)
SMILES string   [H]C(=O)CCCCCCCCC
InChI   1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3
InChI key   KSMVZQYAVGTKIV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
HD6000000
Flash Point(F) 
181.4 °F
Flash Point(C) 
83 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D7384 Decanal, ≥98% (GC), liquid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?