EMAIL THIS PAGE TO A FRIEND
128929 Sigma-Aldrich

Tribenzylamine

≥99%

  • CAS Number 620-40-6

  • Linear Formula (C6H5CH2)3N

  • Molecular Weight 287.40

  •  Beilstein/REAXYS Number 2214682

  •  EC Number 210-638-3

  •  MDL number MFCD00004773

  •  PubChem Substance ID 24847879

Purchase

Properties

assay   ≥99%
mp   91-94 °C (lit.)
SMILES string   C(N(Cc1ccccc1)Cc2ccccc2)c3ccccc3
InChI   1S/C21H21N/c1-4-10-19(11-5-1)16-22(17-20-12-6-2-7-13-20)18-21-14-8-3-9-15-21/h1-15H,16-18H2
InChI key   MXHTZQSKTCCMFG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
399.2 °F
Flash Point(C) 
204 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

90660 Tribenzylamine, ≥99.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?