• USA Home
  • 129062 - α-Chloro-α-hydroxy-o-toluenesulfonic acid γ-sultone

EMAIL THIS PAGE TO A FRIEND
129062 Sigma-Aldrich

α-Chloro-α-hydroxy-o-toluenesulfonic acid γ-sultone

99%

Purchase

Properties

assay   99%
mp   114-116 °C (lit.)
SMILES string   ClC1OS(=O)(=O)c2ccccc12
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?