EMAIL THIS PAGE TO A FRIEND
130133 Sigma-Aldrich

5-Nitroacenaphthene

85%

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, N, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   CUARLQDWYSRQDF-UHFFFAOYSA-N
assay   85%
mp   100-101 °C (lit.)

Description

Other Notes

Contains 3-nitroacenaphthene

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AB1060000

Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Clitoral/Preputial Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Clitoral/Preputial Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Ear Zymbal's Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Mammary Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Ovary Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Clitoral Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Ovary

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?