• USA Home
  • 130842 - 3-Acetoxy-2(1H)-pyridone

EMAIL THIS PAGE TO A FRIEND
130842 Sigma-Aldrich

3-Acetoxy-2(1H)-pyridone

97%

Synonym: 1,2-Dihydro-2-oxo-3-pyridyl acetate

Purchase

Properties

assay   97%
mp   145-149 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?