• USA Home
  • 131288 - Orphenadrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
131288

Orphenadrine hydrochloride

99%

Synonym: N,N-Dimethyl-2-(2-methyl-α-phenylbenzyloxy)ethylamine hydrochloride, N,N-Dimethyl-2-(2-methylbenzhydryloxy)ethylamine hydrochloride

  • CAS Number 341-69-5

  • Empirical Formula (Hill Notation) C18H23NO · HCl

  • Molecular Weight 305.84

  •  Beilstein/REAXYS Number 3745818

  •  MDL number MFCD00012480

Purchase

Properties

assay   99%
SMILES string   Cl.CN(C)CCOC(c1ccccc1)c2ccccc2C
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?