• USA Home
  • 13217 - Manganese(II) chloride

EMAIL THIS PAGE TO A FRIEND
13217 Aldrich

Manganese(II) chloride

purum, anhydrous, 98-102% (AT)

Synonym: Manganese dichloride, Sacchite

Purchase

Properties

InChI Key   GLFNIEUTAYBVOC-UHFFFAOYSA-L
grade   purum
assay   98-102% (AT)
loss   ≤2% loss on drying
mp   652 °C(lit.)
density   2.98 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3288 6.1/PG 3
WGK Germany 
1
RTECS 
OO9625000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

450995 Manganese(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
244589 Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
416479 Manganese(II) chloride, flakes, 97%
328146 Manganese(II) chloride, beads, 98%
429449 Manganese(II) chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?