EMAIL THIS PAGE TO A FRIEND
133000 Sigma-Aldrich

N-Ethylbutylamine

99%

  • CAS Number 13360-63-9

  • Linear Formula CH3CH2CH2CH2NHCH2CH3

  • Molecular Weight 101.19

  •  Beilstein/REAXYS Number 1731324

  •  EC Number 236-415-0

  •  MDL number MFCD00009428

Purchase

Properties

vapor density   3.5 (vs air)
vapor pressure   18 mmHg ( 20 °C)
InChI Key   QHCCDDQKNUYGNC-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.405 (lit.)
bp   108 °C (lit.)
density   0.74 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGII
WGK Germany 
1
RTECS 
EO4880000
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

03205 N-Ethylbutylamine, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?