• USA Home
  • 133183 - 2-Amino-4-imino-2-thiazoline hydrochloride

EMAIL THIS PAGE TO A FRIEND
133183 Aldrich

2-Amino-4-imino-2-thiazoline hydrochloride

99%

  • CAS Number 36518-76-0

  • Empirical Formula (Hill Notation) C3H5N3S · HCl

  • Molecular Weight 151.62

  •  MDL number MFCD00012721

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?