• USA Home
  • 135062 - 5-Bromoindoxyl diacetate

EMAIL THIS PAGE TO A FRIEND
135062 Sigma-Aldrich

5-Bromoindoxyl diacetate

99%

Synonym: 1-Acetyl-5-bromoindol-3-ol acetate

Purchase

Properties

assay   99%
mp   125-128 °C (lit.)
SMILES string   CC(=O)Oc1cn(C(C)=O)c2ccc(Br)cc12
InChI   1S/C12H10BrNO3/c1-7(15)14-6-12(17-8(2)16)10-5-9(13)3-4-11(10)14/h3-6H,1-2H3
InChI key   XJRIDJAGAYGJCK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?