• USA Home
  • 137758 - 4-Chloro-1,2-dimethylbenzene

EMAIL THIS PAGE TO A FRIEND
137758 Sigma-Aldrich

4-Chloro-1,2-dimethylbenzene

technical grade, 90%

Synonym: 3,4-Dimethylchlorobenzene, 4-Chloro-o-xylene

  • CAS Number 615-60-1

  • Linear Formula (CH3)2C6H3Cl

  • Molecular Weight 140.61

  •  Beilstein/REAXYS Number 2076514

  •  EC Number 210-438-6

  •  MDL number MFCD00000596

  •  PubChem Substance ID 24848347

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
grade   technical grade
InChI Key   HNQLMBJUMVLFCF-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.528 (lit.)
bp   221-223 °C(lit.)
density   1.047 g/mL at 25 °C (lit.)

Description

General description

4-Chloro-1,2-dimethylbenzene reacts with various amines, mercaptans and nitriles under aryne forming conditions to yield 4-substituted 1,2-dimethyl benzenes.

Packaging

100 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
Flash Point(F) 
152.6 °F
Flash Point(C) 
67 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?