EMAIL THIS PAGE TO A FRIEND
138002 Sigma-Aldrich

Pentachloropyridine

98%

  • CAS Number 2176-62-7

  • Empirical Formula (Hill Notation) C5Cl5N

  • Molecular Weight 251.33

  •  Beilstein/REAXYS Number 155197

  •  EC Number 218-535-5

  •  MDL number MFCD00006230

  •  PubChem Substance ID 24848226

Purchase

Properties

vapor density   8.4 (vs air)
vapor pressure   0.01 mmHg ( 20 °C)
assay   98%
mp   122-124 °C (lit.)
SMILES string   Clc1nc(Cl)c(Cl)c(Cl)c1Cl
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
UT7000000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?